Assay Design



Gene Transcripts

Gene Symbol
PRELID1
Entrez Gene ID
102185265
Gene Name
PRELI domain containing 1
Gene Aliases
-
Chromosome Location
Chr.7: 71841348 - 71844944 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005682752.3 XP_005682809.1 4 - 5 729 84
GenBank mRNA JR121320.1 - - 101

View Full Product Details