Assay Design
Gene Transcripts
- Gene Symbol
- ZNF24
- Entrez Gene ID
-
102175819
- Gene Name
- zinc finger protein 24
- Gene Aliases
-
-
- Chromosome Location
-
Chr.24: 22022689 - 22033493 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO628909.2 |
- |
2 - 3 |
704 |
82 |
KA305913.1 |
- |
2 - 3 |
484 |
82 |
View Full Product Details