Assay Design



Gene Transcripts

Gene Symbol
NSMCE2
Entrez Gene ID
102186780
Gene Name
NSE2 (MMS21) homolog, SMC5-SMC6 complex SUMO ligase
Gene Aliases
-
Chromosome Location
Chr.14: 66949289 - 67180407 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688848.3 XP_005688905.1 2 - 3 296 98
XM_005688849.3 XP_005688906.1 2 - 3 294 98
GenBank mRNA JT699390.1 - 8 - 9 875 98

View Full Product Details