Assay Design



Gene Transcripts

Gene Symbol
LOC102180151
Entrez Gene ID
102180151
Gene Name
monocarboxylate transporter 13
Gene Aliases
SLC16A13
Chromosome Location
Chr.19: 26628185 - 26631986 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064520.1 XP_017920009.1 2 - 3 763 63
XM_018064522.1 XP_017920011.1 2 - 3 261 63
GenBank mRNA KA312316.1 - 2 - 3 1313 63

View Full Product Details