Assay Design



Gene Transcripts

Gene Symbol
ATP6V1G2
Entrez Gene ID
102187358
Gene Name
ATPase H+ transporting V1 subunit G2
Gene Aliases
-
Chromosome Location
Chr.23: 22218954 - 22221213 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696607.3 XP_005696664.1 2 - 3 378 72
GenBank mRNA JO634396.2 - 2 - 3 430 72

View Full Product Details