Assay Design



Gene Transcripts

Gene Symbol
GATAD1
Entrez Gene ID
102184028
Gene Name
GATA zinc finger domain containing 1
Gene Aliases
-
Chromosome Location
Chr.4: 110827534 - 110839719 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047432.1 XP_017902921.1 2 - 3 584 86
XM_018047433.1 XP_017902922.1 2 - 3 526 86
GenBank mRNA JT665159.1 - 2 - 3 453 86

View Full Product Details