Assay Design



Gene Transcripts

Gene Symbol
PPY
Entrez Gene ID
102179133
Gene Name
pancreatic polypeptide
Gene Aliases
-
Chromosome Location
Chr.19: 43455635 - 43457213 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972519.2 XP_013827973.1 3 - 4 330 85

View Full Product Details