Assay Design



Gene Transcripts

Gene Symbol
MZB1
Entrez Gene ID
102169583
Gene Name
marginal zone B and B1 cell specific protein
Gene Aliases
-
Chromosome Location
Chr.7: 59834328 - 59836444 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005683014.3 XP_005683071.1 2 - 3 415 98
GenBank mRNA JO580795.1 - 2 - 3 507 98

View Full Product Details