Assay Design



Gene Transcripts

Gene Symbol
YARS2
Entrez Gene ID
102179742
Gene Name
tyrosyl-tRNA synthetase 2
Gene Aliases
-
Chromosome Location
Chr.5: 75689579 - 75701977 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680682.3 XP_005680739.1 4 - 5 1359 98

View Full Product Details