Assay Design



Gene Transcripts

Gene Symbol
SYT4
Entrez Gene ID
102170652
Gene Name
synaptotagmin 4
Gene Aliases
-
Chromosome Location
Chr.24: 12775729 - 12785834 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005696966.3 XP_005697023.2 3 - 4 1218 66
GenBank mRNA JT728313.1 - 3 - 4 1145 66

View Full Product Details