Assay Design



Gene Transcripts

Gene Symbol
NUP58
Entrez Gene ID
102168273
Gene Name
nucleoporin 58
Gene Aliases
NUPL1
Chromosome Location
Chr.12: 52830225 - 52862818 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056567.1 XP_017912056.1 9 - 10 1185 70
GenBank mRNA JO351699.2 - 1 - 2 58 70

View Full Product Details