Assay Design



Gene Transcripts

Gene Symbol
PXMP4
Entrez Gene ID
102189771
Gene Name
peroxisomal membrane protein 4
Gene Aliases
-
Chromosome Location
Chr.13: 62730701 - 62747007 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005688471.3 XP_005688528.3 1 - 2 308 97
GenBank mRNA JO437404.2 - 1 - 2 330 97
KA327044.1 - 3 - 4 945 97

View Full Product Details