Assay Design



Gene Transcripts

Gene Symbol
HS6ST3
Entrez Gene ID
102188872
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
-
Chromosome Location
Chr.12: 12248001 - 12966830 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056433.1 XP_017911922.1 1 - 2 923 61
GenBank mRNA JO628433.2 - 1 - 2 93 61

View Full Product Details