Assay Design



Gene Transcripts

Gene Symbol
C5H12orf66
Entrez Gene ID
102182671
Gene Name
chromosome 5 C12orf66 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 48834201 - 48857641 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680225.3 XP_005680282.2 1 - 2 286 86

View Full Product Details