Assay Design



Gene Transcripts

Gene Symbol
C24H18orf54
Entrez Gene ID
102187444
Gene Name
chromosome 24 C18orf54 homolog
Gene Aliases
-
Chromosome Location
Chr.24: 53955279 - 53976927 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039777.1 XP_017895266.1 5 - 6 1573 78

View Full Product Details