Assay Design



Gene Transcripts

Gene Symbol
POU4F2
Entrez Gene ID
102178559
Gene Name
POU class 4 homeobox 2
Gene Aliases
-
Chromosome Location
Chr.17: 59287937 - 59291562 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061378.1 XP_017916867.1 1 - 2 534 51

View Full Product Details