Assay Design



Gene Transcripts

Gene Symbol
PEG10
Entrez Gene ID
102183389
Gene Name
paternally expressed 10
Gene Aliases
-
Chromosome Location
Chr.4: 108562825 - 108575496 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291297.1 NP_001278226.1 1 - 2 221 89
NM_001291298.1 NP_001278227.1 1 - 2 221 89
GenBank mRNA KA339823.1 - 1 - 2 208 89

View Full Product Details