Assay Design



Gene Transcripts

Gene Symbol
LALBA
Entrez Gene ID
100860779
Gene Name
lactalbumin alpha
Gene Aliases
ALA
Chromosome Location
Chr.5: 30812572 - 30814588 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285635.1 NP_001272564.1 2 - 3 307 78
GenBank mRNA EF564266.1 - 2 - 3 307 78
X05149.1 - 2 - 3 305 78

View Full Product Details