Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289948.1 | NP_001276877.1 | 4 - 5 | 576 | 89 |
GenBank mRNA | JO352263.2 | - | 1 - 2 | 353 | 89 |
JO388690.2 | - | 1 - 2 | 367 | 89 | |
JO446296.2 | - | 3 - 4 | 486 | 89 | |
JO523797.2 | - | 1 - 2 | 296 | 89 | |
JO548702.2 | - | 1 - 2 | 299 | 89 | |
JO565982.2 | - | 3 - 4 | 516 | 89 | |
JO586623.2 | - | 1 - 2 | 330 | 89 | |
JT659238.1 | - | 3 - 4 | 89 | ||
JT706680.1 | - | 5 - 6 | 671 | 89 | |
JT716838.1 | - | 5 - 6 | 575 | 89 | |
JT738521.1 | - | 3 - 4 | 516 | 89 | |
KA327599.1 | - | 1 - 2 | 382 | 89 | |
KC537060.1 | - | 4 - 5 | 576 | 89 |