Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001285737.1 | NP_001272666.1 | 1 - 2 | 498 | 66 |
GenBank mRNA | AY436347.1 | - | 1 - 2 | 372 | 66 |
GQ246166.1 | - | 1 - 2 | 498 | 66 | |
GU377303.1 | - | 1 - 2 | 372 | 66 | |
HM462259.1 | - | 1 - 2 | 493 | 66 | |
HM462261.1 | - | 1 - 2 | 493 | 66 | |
JN639006.1 | - | 1 - 2 | 372 | 66 | |
JN662463.1 | - | 1 - 2 | 372 | 66 | |
JX840482.1 | - | 1 - 2 | 372 | 66 |