Assay Design



Gene Transcripts

Gene Symbol
TAC3
Entrez Gene ID
100860865
Gene Name
tachykinin 3
Gene Aliases
GNKB
Chromosome Location
Chr.5: 55735711 - 55742398 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001287572.1 NP_001274501.1 6 - 7 529 86
GenBank mRNA AB796345.1 - 6 - 7 529 86

View Full Product Details