Assay Design
Gene Transcripts
- Gene Symbol
- OPRK1
- Entrez Gene ID
-
102181222
- Gene Name
- opioid receptor kappa 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.14: 60304540 - 60335238 on Build ARS1
- Gene Symbol
- OPRL1
- Entrez Gene ID
-
102177174
- Gene Name
- opioid related nociceptin receptor 1
- Gene Aliases
-
-
- Chromosome Location
-
Chr.13: 53099265 - 53108771 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO622087.2 |
- |
2 - 2 |
2325 |
83 |
View Full Product Details