Assay Design



Gene Transcripts

Gene Symbol
DGAT2
Entrez Gene ID
102174409
Gene Name
diacylglycerol O-acyltransferase 2
Gene Aliases
-
Chromosome Location
Chr.15: 27720878 - 27753735 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001314305.1 NP_001301234.1 1 - 2 49 57
GenBank mRNA HM566448.1 - 1 - 2 50 57
KA327728.1 - 1 - 2 233 57

View Full Product Details