Assay Design



Gene Transcripts

Gene Symbol
GLYCAM1
Entrez Gene ID
100860769
Gene Name
glycosylation-dependent cell adhesion molecule-1
Gene Aliases
GlyCAM-1, PP3
Chromosome Location
Chr.5: 25217299 - 25220218 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285626.1 NP_001272555.1 1 - 2 138 61
GenBank mRNA AJ249734.1 - 1 - 2 138 61
EF564265.1 - 1 - 2 116 61

View Full Product Details