Assay Design



Gene Transcripts

Gene Symbol
IL12A
Entrez Gene ID
100861293
Gene Name
interleukin 12A
Gene Aliases
IL-12A
Chromosome Location
Chr.1: 106965309 - 106971753 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285638.1 NP_001272567.1 1 - 2 169 111
GenBank mRNA AF003542.1 - 1 - 2 169 111
AY603407.1 - 1 - 2 121 111

View Full Product Details