Assay Design



Gene Transcripts

Gene Symbol
FGF2
Entrez Gene ID
102186892
Gene Name
fibroblast growth factor 2
Gene Aliases
-
Chromosome Location
Chr.17: 36227054 - 36278138 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018061205.1 XP_017916694.1 2 - 3 810 100
GenBank mRNA JO514963.2 - 1 - 2 230 100

View Full Product Details