Assay Design



Gene Transcripts

Gene Symbol
SLC25A5
Entrez Gene ID
102173075
Gene Name
solute carrier family 25 member 5
Gene Aliases
-
Chromosome Location
Chr.Un|NW_017189785: 53746 - 56558 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018044743.1 XP_017900232.1 2 - 3 703 99
GenBank mRNA JO581318.1 - 2 - 3 610 99
KA317053.1 - 2 - 3 565 99

View Full Product Details