Assay Design



Gene Transcripts

Gene Symbol
ISG20L2
Entrez Gene ID
102174077
Gene Name
interferon stimulated exonuclease gene 20 like 2
Gene Aliases
-
Chromosome Location
Chr.3: 106003102 - 106008284 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677337.3 XP_005677394.1 2 - 3 956 63
GenBank mRNA JO576763.1 - 2 - 3 1055 63

View Full Product Details