Assay Design
Gene Transcripts
- Gene Symbol
- DHODH
- Entrez Gene ID
-
102175324
- Gene Name
- dihydroorotate dehydrogenase (quinone)
- Gene Aliases
-
-
- Chromosome Location
-
Chr.18: 40276917 - 40289923 on Build ARS1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
JO414849.2 |
- |
7 - 8 |
1355 |
79 |
View Full Product Details