Assay Design



Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
102175324
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
-
Chromosome Location
Chr.18: 40276917 - 40289923 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005692197.3 XP_005692254.2 8 - 9 1203 102
XM_005692201.3 XP_005692258.1 7 - 8 972 102
XM_013971133.2 XP_013826587.2 9 - 10 1299 102
XM_013971134.2 XP_013826588.2 9 - 10 1270 102
GenBank mRNA JO414849.2 - 1 - 2 455 102

View Full Product Details