Assay Design



Gene Transcripts

Gene Symbol
HIRIP3
Entrez Gene ID
102188009
Gene Name
HIRA interacting protein 3
Gene Aliases
-
Chromosome Location
Chr.25: 26178195 - 26181071 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005697676.3 XP_005697733.2 6 - 7 1568 80
GenBank mRNA JT670997.1 - 7 - 8 1983 80

View Full Product Details