Assay Design



Gene Transcripts

Gene Symbol
CLDN8
Entrez Gene ID
102182828
Gene Name
claudin 8
Gene Aliases
-
Chromosome Location
Chr.1: 4114961 - 4117073 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674708.3 XP_005674765.1 1 - 1 818 89
GenBank mRNA JO441682.2 - 1 - 1 254 89

View Full Product Details