Assay Design



Gene Transcripts

Gene Symbol
ZNF18
Entrez Gene ID
102182535
Gene Name
zinc finger protein 18
Gene Aliases
-
Chromosome Location
Chr.19: 30351701 - 30377237 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064668.1 XP_017920157.1 5 - 6 914 81
XM_018064669.1 XP_017920158.1 5 - 6 1066 81
XM_018064670.1 XP_017920159.1 5 - 6 911 81

View Full Product Details