Assay Design



Gene Transcripts

Gene Symbol
IL17REL
Entrez Gene ID
102182289
Gene Name
interleukin 17 receptor E like
Gene Aliases
-
Chromosome Location
Chr.5: 118320952 - 118332778 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048934.1 XP_017904423.1 6 - 7 3093 72
GenBank mRNA JO614510.2 - 7 - 8 649 73

View Full Product Details