Assay Design



Gene Transcripts

Gene Symbol
SACS
Entrez Gene ID
102169306
Gene Name
sacsin molecular chaperone
Gene Aliases
sacsin
Chromosome Location
Chr.12: 52089916 - 52171305 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056555.1 XP_017912044.1 7 - 8 1212 91
XM_018056556.1 XP_017912045.1 7 - 8 1212 91

View Full Product Details