Assay Design



Gene Transcripts

Gene Symbol
TP53I13
Entrez Gene ID
102171006
Gene Name
tumor protein p53 inducible protein 13
Gene Aliases
-
Chromosome Location
Chr.19: 20645696 - 20650438 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018064350.1 XP_017919839.1 4 - 5 483 70
XM_018064351.1 XP_017919840.1 4 - 5 362 70
GenBank mRNA JO432285.2 - 2 - 3 794 70

View Full Product Details