Assay Design



Gene Transcripts

Gene Symbol
GAB2
Entrez Gene ID
102177137
Gene Name
GRB2 associated binding protein 2
Gene Aliases
-
Chromosome Location
Chr.29: 17081590 - 17259527 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018043071.1 XP_017898560.1 4 - 5 1290 67
GenBank mRNA JO578193.1 - 4 - 5 1243 67

View Full Product Details