Assay Design



Gene Transcripts

Gene Symbol
ASF1B
Entrez Gene ID
102188759
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
-
Chromosome Location
Chr.7: 96968876 - 96980284 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013965082.2 XP_013820536.2 2 - 3 518 96
GenBank mRNA JO597765.2 - 2 - 3 559 96
KA328482.1 - 2 - 3 366 96

View Full Product Details