Assay Design



Gene Transcripts

Gene Symbol
ZGLP1
Entrez Gene ID
102187940
Gene Name
zinc finger GATA like protein 1
Gene Aliases
-
Chromosome Location
Chr.7: 94721924 - 94724703 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051673.1 XP_017907162.1 4 - 5 733 69

View Full Product Details