Assay Design



Gene Transcripts

Gene Symbol
CBX8
Entrez Gene ID
102186051
Gene Name
chromobox 8
Gene Aliases
-
Chromosome Location
Chr.19: 51962311 - 51967236 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972499.2 XP_013827953.2 1 - 2 227 71
XM_018063650.1 XP_017919139.1 2 - 3 233 71
GenBank mRNA KA353340.1 - 3 - 4 1203 71

View Full Product Details