Assay Design



Gene Transcripts

Gene Symbol
HSBP1L1
Entrez Gene ID
102187361
Gene Name
heat shock factor binding protein 1 like 1
Gene Aliases
-
Chromosome Location
Chr.24: 420438 - 433614 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018039396.1 XP_017894885.1 2 - 3 534 70
XM_018039397.1 XP_017894886.1 2 - 3 521 70
GenBank mRNA KA345838.1 - 2 - 3 532 70

View Full Product Details