Assay Design



Gene Transcripts

Gene Symbol
WNT8B
Entrez Gene ID
102176193
Gene Name
Wnt family member 8B
Gene Aliases
-
Chromosome Location
Chr.26: 30304224 - 30335372 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018041516.1 XP_017897005.1 5 - 6 631 253

View Full Product Details