Assay Design



Gene Transcripts

Gene Symbol
AURKC
Entrez Gene ID
102172930
Gene Name
aurora kinase C
Gene Aliases
-
Chromosome Location
Chr.18: 65929446 - 65933266 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693002.3 XP_005693059.1 5 - 6 774 117
XM_005693003.3 XP_005693060.1 5 - 6 771 117
GenBank mRNA JQ284043.1 - 1 - 2 158 117

View Full Product Details