Assay Design



Gene Transcripts

Gene Symbol
TMIGD1
Entrez Gene ID
102173034
Gene Name
transmembrane and immunoglobulin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.19: 21218093 - 21232767 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693291.3 XP_005693348.1 1 - 2 175 121
GenBank mRNA JP351172.2 - 5 - 6 849 121

View Full Product Details