Assay Design



Gene Transcripts

Gene Symbol
PIN4
Entrez Gene ID
102185726
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Gene Aliases
-
Chromosome Location
Chr.X|NW_017189516: 9797483 - 9806163 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005700651.2 XP_005700708.1 2 - 3 209 99
GenBank mRNA KA323845.1 - 2 - 3 299 99

View Full Product Details