Assay Design



Gene Transcripts

Gene Symbol
NRIP2
Entrez Gene ID
102170860
Gene Name
nuclear receptor interacting protein 2
Gene Aliases
-
Chromosome Location
Chr.5: 105841674 - 105849795 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018048636.1 XP_017904125.1 3 - 4 730 61

View Full Product Details