Assay Design



Gene Transcripts

Gene Symbol
NUDT16
Entrez Gene ID
102175719
Gene Name
nudix hydrolase 16
Gene Aliases
-
Chromosome Location
Chr.1: 138166502 - 138168103 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018051397.1 XP_017906886.1 2 - 3 457 109
GenBank mRNA JO445059.2 - 1 - 2 456 109
JO453423.2 - 1 - 2 456 109

View Full Product Details