Assay Design



Gene Transcripts

Gene Symbol
DACT1
Entrez Gene ID
102175028
Gene Name
dishevelled binding antagonist of beta catenin 1
Gene Aliases
-
Chromosome Location
Chr.10: 32308152 - 32319070 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054093.1 XP_017909582.1 3 - 4 758 92
GenBank mRNA JT710752.1 - 3 - 4 592 92

View Full Product Details