Assay Design



Gene Transcripts

Gene Symbol
C1H21orf91
Entrez Gene ID
102175802
Gene Name
chromosome 1 C21orf91 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 17836540 - 17868813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674758.3 XP_005674815.2 2 - 3 336 67
XM_018051751.1 XP_017907240.1 2 - 3 231 67
XM_018051818.1 XP_017907307.1 2 - 3 233 67
GenBank mRNA JO329956.2 - 3 - 4 226 67
JO329957.2 - 3 - 4 226 67
JO329958.2 - 3 - 4 226 67
JO532561.2 - 2 - 3 211 67
JO611772.2 - 3 - 4 1143 67
KA341029.1 - 2 - 3 206 67

View Full Product Details