Assay Design



Gene Transcripts

Gene Symbol
C5H12orf10
Entrez Gene ID
102179916
Gene Name
chromosome 5 C12orf10 homolog
Gene Aliases
-
Chromosome Location
Chr.5: 26450649 - 26457598 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018047680.1 XP_017903169.1 3 - 4 540 62
GenBank mRNA JO639605.2 - 4 - 5 727 62

View Full Product Details